Needle holders for specimen loops and dissecting needles
Leverandør: HAMMACHER KARL
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
HWO022-24 HWO024-24
631-7143EA 353 DKK
631-7143 233-0099
Needle holders for specimen loops and dissecting needles
Dissekering og inokulering af loop- og nåleholdere
Needle holders for specimen loops and dissecting needles up to a max. diameter of 1,0 mm.