Dispensers for personal protective equipment
Leverandør: GEBRA
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
4209100 4304100 4201100 4208200 4303100 4214100 4208100 4215100 4102100 4102200 4105100 4105200 4109100 4109200 4201200 4209200 4214200 4312100 4310100 4311100 4303120
111-0001EA 457 DKK
111-0001 111-0004 111-0009 111-3710 111-0017 111-3713 111-3709 111-0024 111-3702 111-3704 111-3706 111-3707 111-3711 111-3712 111-3700 111-3705 111-3714 111-3701 111-3708 111-3703 GEBR4303120
Dispensers for personal protective equipment
PPE-dispensere
These containers are designed for storing personal safety apparatus of all kinds in the workplace.
  • ABS construction
  • Protect against dirt, dust and dampness
  • Patented opening allows for quick removal of the PPE in the case of an emergency
Bestillingsoplysninger: Fås i 3 størrelser.