Farveskåle
Leverandør: GLASWARENFABRIK KARL HECHT
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
41205001 41205002 41205010
631-0347EA 326 DKK
631-0347 630-2600 631-0346
Farveskåle
Dåser Farvekrukker
Soda-lime glass.
  • Removable cover
Staining dish for 76×26 mm slides or slides with width up to 45 mm.