Petri dishes, Sterilin®
Leverandør: STERILIN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
124-17 122-17 123 121V PF55 PF55V
391-2022EA 3220 DKK
391-2022 391-2020 391-2021 391-2019 391-2041 391-2042
Petri dishes, Sterilin®
Dyrkningsplader Petriskåle
Ideal for use when savings in media or incubator space are required.
  • Aseptically manufactured under cleanroom conditions (Class 7 ISO 14644) to exclude microbiological contamination
  • 55 mm dish accommodates 47 mm membrane filters making it suitable for water testing
  • 50 mm deep form dish is over 20 mm deep and designed for use with liquid media
Dimensions shown are for outer Ø base and outer height (mm).
Save incubator space with Thermo Scientific™ Sterilin™ 30 to 140 mm Petri Dishes. These small dishes are suitable for most applications. Not available in America.
140 mm dish is produced and tested in accordance with the EN ISO 24998 Petri dish standard which includes stringent dimensional controls.