Petriskåle, Steriplan®, DURAN®
Leverandør: DWK Life Sciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
237554008 237554205 237554505 237554608 237554805 237555107 237555201 237555604 237555904 237553903
391-2810EA 28.3 DKK
391-2810 391-2820 391-2831 391-2830 391-2840 391-2850 391-2860 391-2870 391-2880 391-2805
Petriskåle, Steriplan®, DURAN®
Dyrkningsplader Petriskåle
Soda-lime glass. Hydrolytic class III. Diameter quoted in the table is the external diameter of the lid. The height refers to the height of just the base.