Instrument trays with knob lid
Leverandør: MUEHLACKER METALWARENFABRIK
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
00108 00116 00124 00132 00160 00168
216-1381EA 368 DKK
216-1381 216-1382 216-1383 216-1384 216-1386 216-1387
Instrument trays with knob lid
Bakker/Tallerkener/Potter Instrumentbakker/-skåle
Rustfrit stål.
  • Smooth surfaces for fast, easy cleaning
  • Satin brushed
  • Good chemical resistance, very good thermal resistance