Evaporating dishes
Leverandør: TECHNICAL QUARTZ SOLUTIONS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
73011 73013 73014
216-0293EA 1320 DKK
216-0293 216-0294 216-0295
Evaporating dishes
Bakker/Tallerkener/Potter Fordampningsbassiner/skåle
Made of silica.
  • Very good chemical resistance
  • Very good temperature resistance
  • Easy to clean