VWR®, Dishes, Evaporating, Shallow
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
216-0557EA 470 DKK
216-0557 216-0562 216-0561 216-0560 216-0558 216-0556 216-0555 216-0559
VWR®, Dishes, Evaporating, Shallow
Bakker/Tallerkener/Potter Fordampningsbassiner/skåle
Porcelæn, glaseret.
  • Very good chemical resistance
  • Heat resistant up to +1000 °C
  • Low thermal expansion
The economical alternative for a wide range of laboratory applications. With spout and flat bottom. Glazed white except for the outside bottom surface.
DIN 12903
Certificeringer: DIN 12903