Basins, silica Vitreosil®
Leverandør: TECHNICAL QUARTZ SOLUTIONS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
73016 73018 73019 73021 73022 73023 73024 73025 73026
216-4473EA 760 DKK
216-4473 216-0296 216-0297 216-4475 216-0298 216-0299 216-0300 TSLA73025 216-0301
Basins, silica Vitreosil®
Bakker/Tallerkener/Potter Fordampningsbassiner/skåle
Basins (capsules) with flat bottom.
  • Chemically Inert
  • Can be used at temperatures in excess of 1000 °C
  • Thermal shock resistant and high electrical resistivity