Skåle
Leverandør: RSG
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
705.180 705.240 705.260 705.330 705.360 705.410 705.300 705.150 705.000
216-1241EA 151 DKK
216-1241 216-1242 216-1243 216-1245 216-1246 216-1247 216-2023 216-2022 216-1249
Skåle
Bakker/Tallerkener/Potter Laboratoriebakker/-skåle
Stainless steel, ¹⁸/₁₀.
  • With folded edge; fast, easy cleaning
  • Polished electrolytically, anti-magnetic
  • Good chemical resistance, very good thermal resistance
Round multipurpose dishes for a wide range of uses.
Bestillingsoplysninger: Each dish set (Cat. No. 216-1249) comprises one of each 1, 2 and 3 litre dishes.