Dilatometers, Quickfit®
Leverandør: SciLabware
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
SB14DL WE12/1
BIBBSB14DLEA 1810 DKK
BIBBSB14DL BIBBWE12/1
Dilatometers, Quickfit®
Dilatometre
PYREX®, borosilicate glass.
  • Dilatometer body is graduated with an arbitrary scale
  • Interchangable stopper has open top to allow weighting with steel balls etc.
For measuring the expansion of edible oils and fats over the temperature range within which melting takes place.
Bestillingsoplysninger: The body and stopper should be ordered separately.