Crucibles, low form
Leverandør: TECHNICAL QUARTZ SOLUTIONS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
73056 73058 73060 73061 73063 73065 73067
459-0237EA 828 DKK
459-0237 216-4479 216-4481 459-0176 459-0172 459-0178 459-0173
Crucibles, low form
Digler
These crucibles are made from fused quartz, translucent, glazed.
  • Max. temperature: 1050 °C
Bestillingsoplysninger: Crucible and lid have to be ordered separately.