Sample containers, with screw cap, STERILIN®
Leverandør: STERILIN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
128DB/IRR 185DB/IRR 190DB/IRR
216-1557EA 6340 DKK
216-1557 216-2129 216-2130
Sample containers, with screw cap, STERILIN®
Prøvecontainere Allround kasser
Polystyrene, clear, with screw cap.
  • Triple-wrapped with two easy tear bags within a bulk outer liner bag
  • Sterilized by gamma irradiation
  • Marked with an irradiation dot as assurance of sterility and batch details for full traceability
  • Leak-tested in accordance with BS EN 14254 Annexe D
Ideal for use in hospital theatres to facilitate a sterile transfer of product.
Certificeringer: CE marking denotes compliance with the European Directive 98/79/EC.
Advarsel: *Produktet/produkterne på denne side er uden for anvendelsesområdet for EU -direktiv 2019/904.