Jars with push-on lid
Leverandør: Brand
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
61815 61820
216-3184EA 240 DKK
216-3184 216-3188
Jars with push-on lid
Dåser Dåser, allround
PP, gennemskinnelig.
  • Very good chemical resistance
  • Heat resistant from −10 to +125 °C
  • Autoclavable
Ideal for sampling, transportation and storage.