Transport and storage containers with snap-on lids
Leverandør: hünersdorff GmbH
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
911100 911200 911300 911400 911600
216-3212EA 0 DKK
216-3212 216-3214 216-3216 216-3218 216-3219
Transport and storage containers with snap-on lids
Prøvecontainere Allround kasser
White HDPE jar with white LDPE lid
  • Good thermal and chemical resistance
  • Freezable, stackable
  • With tightly sealing lid
For storing and transporting solid and liquid materials, samples and general labware.