Jars with screw cap
Leverandør: DELTALAB
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
202841 202842 202844 202840 202843 202848
216-4301EA 2160 DKK
216-4301 216-4302 216-4303 216-2056 216-2057 216-2058
Jars with screw cap
Dåser Dåser, allround
White PP with yellow safety screw cap.
  • Excellent thermal and chemical resistance
  • Graduated
  • Airtight, liquid-tight
Ideal for transporting and storing biological materials, liquids, solutions and powdery materials.