Adaptere med rørtilslutning
Leverandør: Lenz Laborglas GmbH & CO.KG
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
05092001 05092003 05092004 05095001 05095002 05095003 05095004
201-2141EA 110 DKK
201-2141 201-0348 201-2142 201-2146 201-0349 201-2109 201-2147
Adaptere med rørtilslutning
Adapter Slangekonnektorer med NS-slib
DURAN®, borosilicate glass, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Adapter with standard ground joint cone and straight or right angled tube connector. The connector has an external diameter of 8 mm.