Slangekoblinger, lige
Leverandør: KARTELL
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
70510 70511 70512 70513 70514 70515
229-0713EA 89.6 DKK
229-0713 229-0714 229-0715 229-0716 229-0717 229-0718
Slangekoblinger, lige
Fittings og stik
PP, hvid
  • Good mechanical strength and chemical resistance
  • Very good thermal resistance
  • Autoclavable
  • Conforms to Food and Drug regulations 90/128 CEE, 92/39 CEE and FDA 21 CFR 177.1520
With tapered ends, for connecting tubing with the same diameter.