Slangekoblinger, Y-formede
Leverandør: KARTELL
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
70464 70465 70466 70467 70468 70525 70526
229-0708EA 95.3 DKK
229-0708 229-0709 229-0710 229-0711 229-0712 229-0721 229-0722
Slangekoblinger, Y-formede
Fittings og stik
PP, gennemskinnelig.
  • Good mechanical strength and chemical resistance
  • Very good thermal resistance
  • Autoclavable
  • Suitable for foodstuff
Dimensions are shown as both nominal and exact as elasticity of tubing will influence the choice of size. Use our tubing selector 229-0111 to determine 'grip'. This will then give equivalent crest dimension for the particular connector concerned.
They meet with the Food and Drug regulations.90/128CEE 92/39CEE and FDA 21 CFR 177.1520.
Suitable for foodstuff.
Certificeringer: Conforms to Food and DrugMore Product Information