Liebig-kondenser med standardslib
Leverandør: Lenz Laborglas GmbH & CO.KG
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
05210311 05210314 05210414 05210312 05210313 05210212 05210411 05210412 05210413 05210514 05210714 05210211
201-1676EA 474 DKK
201-1676 201-1675 201-1684 201-1170 201-0903 201-1171 LENZ05210411 LENZ05210412 LENZ05210413 LENZ05210514 LENZ05210714 201-0902
Liebig-kondenser med standardslib
Svaler
DURAN®, borosilicate 3.3 glass, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
For condensing vapours, with fused mantle, cone and socket. Water supplied through plastic connectors with 8 mm external diameter and GL 14 threaded joints.
DIN 12576
Certificeringer: DIN 12576.