Dimroth condensers with standard ground joints
Leverandør: Lenz Laborglas GmbH & CO.KG
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
05240201 05240302 05240303 05240304 05240404 05240202 05240301 05240406 05240306
201-3331EA 825 DKK
201-3331 201-0362 201-0363 201-3332 201-3333 LENZ05240202 LENZ05240301 LENZ05240406 LENZ05240306
Dimroth condensers with standard ground joints
Svaler
DURAN®, borosilikatglas, klart.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
For cooling condensate from vapours, with core and jacket. Water supplied through glass connectors with 8 mm external.