Dimroth kondenser med standard-slib
Leverandør: Lenz Laborglas GmbH & CO.KG
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
05240211 05240314 05240414 05240311 05240212 05240312 05240313 05240316 05240416
201-1671EA 993 DKK
201-1671 201-1672 201-1673 LENZ05240311 LENZ05240212 LENZ05240312 LENZ05240313 LENZ05240316 LENZ05240416
Dimroth kondenser med standard-slib
Svaler
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
For condensing vapours. Water supplied through plastic connectors with 8 mm external diameter and GL 14 threaded joints.