Cation exchange HPLC columns, PRP-X400
Leverandør: HAMILTON BONADUZ
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
79398 79717 79473 79387 79131 79132 79376 79377 79225 79224 79290 79916
554-0226EA 9480 DKK
554-0226 554-0402 554-0253 554-0218 554-1231 554-1232 554-1251 554-1252 554-2535 554-2534 HAMI79290 HAMI79916
Cation exchange HPLC columns, PRP-X400
Kromatografikolonner
Polymeric cation exchange packing for separation of glyphosate and its metabolite in drinking water.
  • Faster analysis compared to other columns (PRP-X200)
  • Detection levels <10 ppb when using the post column, OPA derivatisation technique