High pressure in-line filters
Leverandør: VICI JOUR RESEARCH
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
JR-M68230-2 JR-M68231-5
554-1156EA 5140 DKK
554-1156 554-1157
High pressure in-line filters
Filtre Indsatsfiltre
The stainless steel in-line filter has virtually no dead volume and can be used in various applications without significant band broadening or loss of efficiency. This version is intended for analytical use between the autosampler injection valve and the column.
  • Minimises dead volume
  • For high pressure applications (up to 350 bar/5000 psi)