VWR® Hichrom Fittings
Leverandør: Avantor
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
HI-050EA 194 DKK
HI-050 HI-050X HI-031
VWR® Hichrom Fittings
Fittings og stik
PEEK. Can be used at temperatures up to 150 °C. Unlike stainless steel fittings, PEEK fingertight fittings will not permanently lock into place on the tubing. Compatible with all types of end fittings.
  • Fingertight to 6000 psi
  • Void-free connection
  • Inert and biocompatible