Fractionating columns, Vigreux, Quickfit®
Leverandør: SciLabware
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
475202 FC3/13 FC3/14 FC3/23
201-8243EA 0 DKK
201-8243 201-8392 BIBBFC3/14 BIBBFC3/23
Fractionating columns, Vigreux, Quickfit®
Destillations komponenter Destillationskolonner
PYREX®, borosilicate glass
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
No glass jacket. With combination of horizontal and inclined indentations. Horizontal indentations ensure intimate vapour and piquid contatc. Inclined indentations redistribute liquid from walls to centre of column.