Chemical resistant overalls, Tychem® 2000 C, models CHA5 / CHA6
Leverandør: DuPont
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
TCCHA5TYLLG002 TCCHA5TYLMD002 TCCHA5TYLSM002 TCCHA5TYLXL002 TYCCHA5TYL16/L TYCCHA5TYL16/M TYCCHA5TYL16/S TCCHA5TYL2X002 CHA5TYL16/XL CHA5TYL16/XXL CHA5TYL16/XXXL TCCHA5TYL3X002
113-0495EA 7840 DKK
113-0495 113-0496 113-0497 113-0498 113-0499 113-0529 113-0530 113-0485 113-0486 113-0487 113-0488 113-3891
Chemical resistant overalls, Tychem® 2000 C, models CHA5 / CHA6
Engangsbeklædning
Made from a lightweight and durable fabric (<450 g per garment), Tychem® 2000 C garments utilise the strength of Tyvek®, and a polymeric barrier coating, to offer good permeation barrier protection against a wide range of inorganic chemicals and biological hazards (even under pressure).
  • Hood; elasticated face, wrists, waist and ankles for an optimal fit
  • Thumb loops prevent sleeves from riding up
  • Self-adhesive double-zip flap and chin flap for tight seal of suit to face masks
  • Stitched, over-taped seams with barrier tape for protection and strength
  • Bright yellow colour for high visibility
  • Anti-static treatment (EN 1149-5) on inside
  • CHA6: With integrated socks to be worn inside shoes or boots and additional knee-length boot flap
Tychem® 2000 C is used for splash or pressurised splash protection in a variety of industrial environments, including pulp and paper manufacturing, food processing, chemical processing and pharmaceutical manufacturing.
Colour: Yellow
Weight: 83 g/m²
Certificeringer: Type 3-B/4-B/5-B/6-B, EN 1149-5, EN 1073-2, EN 14126; Cat. III.; (EU) 2016/425
Emballering: 25 per box (without socks) and 20 per box (with socks), bulk packed.