Cleanroom face veils, Kimtech™ M7, PE
Leverandør: KIMBERLY CLARK
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
62757 62751
111-0384EA 867 DKK
111-0384 113-0929
Cleanroom face veils, Kimtech™ M7, PE
Masker Renrumsmasker
Single-use personal veil designed to fit most garment systems.
  • Made from low linting breathable PE film
  • Knitted headbands or snap fasteners
  • Intended for use in cleanrooms where reusable face veils are currently worn
Suitable for cleanrooms ISO class 7 or higher.
Emballering: Pakket med 25 kapper pr. pose, 6 poser pr. kasse.