Ekstra fittings til hurtig påfyldnings/ventileringslåg, Nalgene®
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
2159-0001 2159-0002 2159-0011 2159-0012
216-4389EA 636 DKK
216-4389 216-4390 216-4391 216-4392
Ekstra fittings til hurtig påfyldnings/ventileringslåg, Nalgene®
Låg Flaskelåg
PP plug connectors with hose barb.
  • Very good chemical resistance, heat resistant up to 135 °C
  • Available in straight or right-angled versions
  • Simple easy assembly and disassembly
Sizes are not interchangeable.