Carboys, wide neck, rectangular, HDPE
Leverandør: MELLERUD PLAST EMBALLATOR
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
48C1000100117 48C0500100117
215-3063EA 211 DKK
215-3063 215-3061
Carboys, wide neck, rectangular, HDPE
Flasker Flasker, dunke
HDPE, natural, without screw cap.
  • Rectangular, save spaces
  • Good chemical resistance
  • Screw cap must be ordered separately