Flasker, smal hals, med tråd
Leverandør: GRAVIS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
C00827 C00834 C00844 C00847 C00854 C00727 C00734 C00744 C00747 C00754 C00764 C00856 C00848 C00873 C00874 C00772124 C00864-35 C00841-130 C00827P09028 C00756 C00748188 C00872124 C00741
215-2613EA 838 DKK
215-2613 215-2614 215-2616 215-2617 215-2618 215-2604 215-2605 215-2608 215-2609 215-2610 215-2611 215-2396 215-2395 215-1886 215-3232 215-2384 215-2397 215-3231 SPVAC00827P09028 215-3230 SPVAC00748188 SPVAC00872124 SPVAC00741
Flasker, smal hals, med tråd
Flasker Glasflasker
Glass, clear, amber or yellow.
  • Neck Ø: 28 mm
  • Good chemical and temperature resistance
  • Amber coloured to protect light sensitive substances
Choice of various screw caps.
Bestillingsoplysninger: Supplied without screw caps - please order separately.
Emballering: Packaged in impermeable plastic packs.