Replacement screw caps
Leverandør: RIXIUS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
8-1000-45 8-1000-68
RIXI8-1000-45EA 128 DKK
RIXI8-1000-45 RIXI8-1000-68
Replacement screw caps
Låg Flaskelåg
PP, black.
  • Very good chemical and thermal resistance
  • Suitable for bottles 215-1821 to 215-1836 and culture bottle 215-1861 to 215-1866
  • LUP lining for better sealing