Bottles, narrow neck, aluminium, with thread, CHEM 42™
Leverandør: TOURNAIRE-EMBALLAGE
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1434S1 1447B1 1447S1 6440S1
215-3857EA 75.6 DKK
215-3857 215-3855 TOUR1447S1 TOUR6440S1
Bottles, narrow neck, aluminium, with thread, CHEM 42™
Flasker Metalflasker
Pure aluminium (99,5%).
  • High mechanical performance
  • Perfect chemical
  • Totally recyclable
Suitable for storage and transport of highly aggessive liquids.