Screw caps for bottles with narrow neck, 301 and 302 series
Leverandør: Kautex
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
801-90605 801-90606 2000090604
KAUT801-90605EA 318 DKK
KAUT801-90605 KAUT801-90606 KAUT2000090604
Screw caps for bottles with narrow neck, 301 and 302 series
Låg Flaskelåg
LDPE, gennemskinnelig
  • Meget god kemisk og termisk resistens
  • Sikret mod lækage
For narrow neck bottles in the 301 and 302 series.