Spencer wells clamps
Leverandør: MURRAY S
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
N518/B1 N518/B2 N518/B3
232-0040EA 199 DKK
232-0040 232-0041 232-0042
Spencer wells clamps
Klemmer Rørklemmer
Stainless steel, rust-proof. For stopping flow in tube connections. Straight with box joint.
  • Very good thermal resistance
  • Easy cleaning
  • Safe, simple operation