Pean clamps
Leverandør: BOCHEM
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
2291 2292 2293
229-0004EA 129 DKK
229-0004 229-0005 229-0006
Pean clamps
Klemmer Rørklemmer
Stainless steel. For stopping flow in tube connections.
  • Very good thermal resistance
  • Easy cleaning
  • Safe, simple operation