Mohr pinch cocks
Leverandør: USBECK Laborgeräte
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
5530 5531 5532 5533 5534
229-0331EA 72.6 DKK
229-0331 229-0332 229-0333 229-0334 229-0335
Mohr pinch cocks
Klemmer Rørklemmer
Nickel-plated brass. For regulating or stopping the flow of liquids in tube connections.
  • Very good thermal resistance
  • Easy handling
  • Easy cleaning