Fork clamps for spherical joints
Leverandør: Lenz Laborglas GmbH & CO.KG
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
01232915 01233520 01234125 01236540 01235130
201-7343EA 162 DKK
201-7343 201-7344 271-7345 LENZ01236540 LENZ01235130
Fork clamps for spherical joints
Klemmer Klemmer til NS-slib
Clips for spherical joints made from stainless steel chrome/nickel.
  • Easy to install and remove
  • Excellent heat and chemical resistance
  • With set screw