Clamp with chain
Leverandør: BOCHEM
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
5424 5485
241-7411EA 1070 DKK
241-7411 BOCH5485
Clamp with chain
Klemmer Universalklemmer
For attaching glass beakers, flasks, reaction vessels and other laboratory vessels to support stands.
  • Adjustable chain with silicone coating, 70 cm
  • Without bosshead for fixing to rod