Clamps with bosshead
Leverandør: USBECK Laborgeräte
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
2640 2650 2660
241-0258EA 176 DKK
241-0258 241-0259 241-0260
Clamps with bosshead
Klemmer Universalklemmer
Zinc die-cast, powder-coated.
  • Cork lined jaws
  • Both jaws are curved