Inoculating loops and needles
Leverandør: KARTELL
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
2650 2651
KART2650EA 1840 DKK
KART2650 KART2651
Inoculating loops and needles
Spredere Podenåle og Loops
High impact PS, sterile.
  • Choice of either 1 µl or 10 µl loop on one end and needle at the other
  • Sterilised by gamma irradiation
Emballering: 20 per bag.