Bacterial cell spreaders, SP Bel-Art
Leverandør: Bel-Art Products, a Part of SP
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
F377360009 F377360006
391-2007EA 378 DKK
391-2007 391-2006
Bacterial cell spreaders, SP Bel-Art
Spredere Celle Spredere
For spreading cells onto the surface of an agar plate to form individual colonies. These stainless spreaders have a smooth triangular end with a straight spreading bar.
  • The small size is ideal for 6 cm plates and the large size for 9 cm plates
  • Autoclavable and may be flame sterilised
Overall length: 203 mm