Burettes, straight stopcock, Schellbach
Leverandør: HIRSCHMANN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
3110160 3110170 3110175 3110260 3110270 3110275
612-3533EA 605 DKK
612-3533 612-3534 612-3536 612-3537 612-3538 612-3539
Burettes, straight stopcock, Schellbach
Buretter
DURAN®, klasse B.
  • Calibrated to deliver 'Ex'
  • Short marks at major graduations, blue graduations
Skalaens længde: 500 mm.
Scale length: 500 mm
Certificeringer: DIN ISO EN 385