Buretter, pelletmønster, laterale og mellemliggende stophaner, Schellbach
Leverandør: HIRSCHMANN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
354016060 354017060 354017560 354037060 354037560
612-4097EA 2260 DKK
612-4097 612-4098 612-4099 612-4101 612-4103
Buretter, pelletmønster, laterale og mellemliggende stophaner, Schellbach
Buretter
DURAN®, klasse AS.
  • Calibrated to deliver 'Ex', conformity certified (DE-M marking) with dated batch identification
  • Schellbach stripe, ring marks at major graduations, blue graduations
  • Automatic zeroing, glass spindle and with intermediate stopcock with glass key or PTFE spindle
DIN EN ISO 385
Certificeringer: DIN EN ISO 385