Buckets with lid, Nalgene®
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
7102-0080 7102-0140
216-0096EA 534 DKK
216-0096 216-0097
Buckets with lid, Nalgene®
Spande
LDPE.
  • Suitable for storage of specimens in formalin
Graduated buckets with nylon-coated wire handle and air tight white lids.