Slide boxes, SlideFile™
Leverandør: Simport Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
M700-100B M700-100G M700-100P M700-100W M700-100Y
631-1900EA 2140 DKK
631-1900 631-1901 631-1902 631-1903 631-1904
Slide boxes, SlideFile™
Æsker til objektglas
This impact resistant storage system can hold up to 400 slides per unit, for space efficient storage. All slides are stored upright for easier manipulation.
  • Removable tray inside the storage box for 100 individual numbered slots
  • Transparent hinged cover makes it easy to see the contents of the box at a glance
  • Unique feature with this system to read barcodes without having to remove the slides from the box
W×D×H: 82×245×86 mm
Bestillingsoplysninger: Each system includes a slide box and a removable tray.
Leveringsinformation: Der leveres indekskort nummeret fra 1 til 100 til registrering af objektglasnavne med hver boks.