Storage Boxes for Cryogenic Tubes
Leverandør: Brand
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
114862 114864 114866
479-1232EA 724 DKK
479-1232 479-1233 479-1234
Storage Boxes for Cryogenic Tubes
Bokse Æsker til Kryorør
PC. The transparent cover allows the user to see the contents of the box.
  • Operating range −196 to +121 °C (in the gaseous phase of liquid nitrogen)
  • Autoclavable at 121 °C (2 bar), acc. DIN EN 285
  • Stackable
Printed with a series of squares (numbered from 1 to 81 or 1 to 100). The surface accepts writing with markers, facilitating inventory control.