Bottles, wide neck, square, with screw caps, Azlon®
Leverandør: SciLabware
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BGE242P BGE240P BGE236P
215-0447EA 871 DKK
215-0447 215-0446 215-0445
Bottles, wide neck, square, with screw caps, Azlon®
Flasker Plastflasker
HDPE, translucent, with plug seal and blue screw cap.
  • Good chemical resistance
  • Leakproof
  • Space saving and lightweight
Perfect for powders and granules. The wide mouth allows easy filling and cleaning. Moulded graduations for approximate volume measurement.