Bottles, wide neck, with screw cap, Azlon®
Leverandør: SciLabware
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BWH0060PN BLH0150P BWH0250PN BWH0500PN BWH1000PN BWH2000P BLH0100P BLH0125P BWH0030PN BLH0100BULK BLH0125BULK BWH0250BULK BWH0500BULK BWH1000BULK BWH2000BULK
215-0540EA 127 DKK
215-0540 215-0389 215-0542 215-0544 215-0546 215-0547 215-3688 215-0518 215-0538 215-4073 215-4074 215-4076 215-4077 215-4078 215-4079
Bottles, wide neck, with screw cap, Azlon®
Flasker Plastflasker
HDPE, gennemskinnelige flasker, stive, med PP skruelåg.
  • Leakproof, linerless design
  • Smooth inner contours for efficient pouring and ease of cleaning
  • Heavy-duty and durable
  • Good chemical resistance
Ideal for a wide range of liquid storage, media preparation and sampling applications. Wide opening allows rapid filling and dispensing of powders, granules and pastes.