Bottles, wide neck, square, with screw cap, series 357
Leverandør: Kautex
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
357-70546 357-70547
215-3207EA 284 DKK
215-3207 215-3208
Bottles, wide neck, square, with screw cap, series 357
Flasker Plastflasker
HDPE, polygonal, natural colour, with screwed-on closure.
  • Wide neck opening
  • Easy to fill, easy to clean
  • Suitable for food
  • Not leakproof
  • Halal certified
The wide neck opening allows for easy filling, emptying, and cleaning. Particularly suitable for storage, transport, or disposal of powdery or granulated materials.